Assay Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133900.4 | NP_598661.1 | 4 - 5 | 758 | 91 | |
| GenBank mRNA | AK088865.1 | - | 4 - 5 | 710 | 91 | |
| AK168048.1 | - | 2 - 3 | 366 | 91 | ||
| BQ177662.1 | - | 2 - 3 | 324 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psph
Genomic Map