Assay Details
| Gene Symbol: Prkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008926.4 | NP_032952.3 | 11 - 12 | 1571 | 94 | |
| GenBank mRNA | AK038739.1 | - | 11 - 12 | 1555 | 94 | |
| AK081422.1 | - | 11 - 12 | 1572 | 94 | ||
| BC113205.1 | - | 11 - 12 | 1482 | 40047751 | 94 | |
| L12460.1 | - | 11 - 12 | 1843 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkg2
Genomic Map