Assay Details
| Gene Symbol: Prkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008926.4 | NP_032952.3 | 6 - 7 | 1082 | 120 | |
| GenBank mRNA | AK038739.1 | - | 6 - 7 | 1066 | 120 | |
| AK081422.1 | - | 6 - 7 | 1083 | 120 | ||
| BC113205.1 | - | 6 - 7 | 993 | 40047751 | 120 | |
| L12460.1 | - | 6 - 7 | 1354 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkg2
Genomic Map