Assay Details
| Gene Symbol: Papss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201470.1 | NP_001188399.1 | 7 - 8 | 999 | 70 | |
| GenBank mRNA | AF085144.1 | - | 7 - 8 | 931 | 70 | |
| AK137675.1 | - | 7 - 8 | 925 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papss2
Genomic Map