Assay Details
| Gene Symbol: Papss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201470.1 | NP_001188399.1 | 9 - 10 | 1341 | 72 | |
| NM_011864.3 | NP_035994.2 | 10 - 11 | 1377 | 72 | ||
| GenBank mRNA | AF052453.1 | - | 10 - 11 | 1358 | 72 | |
| AK128962.1 | - | 9 - 10 | 1362 | 72 | ||
| AK137675.1 | - | 9 - 10 | 1267 | 72 | ||
| BC090997.1 | - | 10 - 11 | 1377 | 30121331 | 72 | |
More Information
Set Membership: |
|

Papss2
Genomic Map