Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 10 - 11 | 1260 | 85 | |
| GenBank mRNA | AB037111.1 | - | 11 - 12 | 1545 | 85 | |
| AB037181.1 | - | 10 - 11 | 1260 | 85 | ||
| AK034493.1 | - | 10 - 11 | 1491 | 85 | ||
| AK046540.1 | - | 10 - 11 | 1454 | 85 | ||
| AK047692.1 | - | 10 - 11 | 1448 | 85 | ||
| AK080951.1 | - | 10 - 11 | 1437 | 85 | ||
| AK166100.1 | - | 10 - 11 | 1508 | 85 | ||
| BC022604.1 | - | 1 - 2 | 42 | 85 | ||
| BC137898.1 | - | 10 - 11 | 1243 | 8860917 | 85 | |
| BC145016.1 | - | 10 - 11 | 1246 | 9053564 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah2
Genomic Map