Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 15 - 16 | 1776 | 79 | |
| GenBank mRNA | AB037111.1 | - | 16 - 17 | 2061 | 79 | |
| AB037181.1 | - | 15 - 16 | 1776 | 79 | ||
| AK080951.1 | - | 15 - 16 | 1953 | 79 | ||
| BC022604.1 | - | 6 - 7 | 558 | 79 | ||
| BC137898.1 | - | 15 - 16 | 1759 | 8860917 | 79 | |
| BC145016.1 | - | 15 - 16 | 1759 | 9053564 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah2
Genomic Map