Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 2 - 3 | 199 | 66 | |
| GenBank mRNA | AB037181.1 | - | 2 - 3 | 199 | 66 | |
| AK034493.1 | - | 2 - 3 | 431 | 66 | ||
| AK047692.1 | - | 2 - 3 | 387 | 66 | ||
| BC137898.1 | - | 2 - 3 | 182 | 8860917 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB037181.1, AK047692.1, BC137898.1, NM_018830.1 transcripts.
Set Membership: |
|

Asah2
Genomic Map