Assay Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037366.1 | - | 1 - 2 | 85 | 109 | |
| AK030601.1 | - | 1 - 2 | 111 | 109 | ||
| BC011278.1 | - | 1 - 2 | 96 | 3674983 | 109 | |
| CN696192.1 | - | 1 - 2 | 134 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF037366.1, AK030601.1, BC011278.1 transcripts.
Set Membership: |
|

Entpd1
Genomic Map