Assay Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008528.4 | NP_032554.2 | 10 - 11 | 1260 | 84 | |
| GenBank mRNA | AB015290.1 | - | 10 - 11 | 934 | 84 | |
| AF068182.1 | - | 10 - 11 | 915 | 84 | ||
| AJ222814.1 | - | 10 - 11 | 887 | 84 | ||
| AK089704.1 | - | 10 - 11 | 901 | 84 | ||
| BC059785.1 | - | 10 - 11 | 886 | 5339723 | 84 | |
| Y17159.1 | - | 10 - 11 | 1260 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Blnk
Genomic Map