Assay Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008528.4 | NP_032554.2 | 1 - 2 | 523 | 147 | |
| GenBank mRNA | AB015290.1 | - | 1 - 2 | 197 | 147 | |
| AF068182.1 | - | 1 - 2 | 178 | 147 | ||
| AJ222814.1 | - | 1 - 2 | 148 | 147 | ||
| AK089704.1 | - | 1 - 2 | 163 | 147 | ||
| AK172016.1 | - | 1 - 2 | 150 | 147 | ||
| BC059785.1 | - | 1 - 2 | 149 | 5339723 | 147 | |
| Y17159.1 | - | 1 - 2 | 523 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB015290.1, AF068182.1, BC059785.1, NM_008528.4, Y17159.1 transcripts.
Set Membership: |
|

Blnk
Genomic Map