Assay Details
| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293757.1 | NP_001280686.1 | 5 - 6 | 665 | 97 | |
| NM_001293758.1 | NP_001280687.1 | 4 - 5 | 498 | 97 | ||
| NM_001293759.1 | NP_001280688.1 | 5 - 6 | 735 | 97 | ||
| NM_011722.4 | NP_035852.1 | 4 - 5 | 552 | 97 | ||
| GenBank mRNA | AF124788.1 | - | 4 - 5 | 283 | 97 | |
| AF190796.1 | - | 4 - 5 | 293 | 97 | ||
| AK003618.1 | - | 4 - 5 | 292 | 97 | ||
| AK088312.1 | - | 4 - 5 | 299 | 97 | ||
| BC029249.1 | - | 4 - 5 | 293 | 4486579 | 97 | |
| CV560309.1 | - | 4 - 5 | 449 | 97 | ||
| DV042569.1 | - | 4 - 5 | 507 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293759.1 transcripts.
Set Membership: |
|

Dctn6
Genomic Map