Assay Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016692.3 | NP_057901.2 | 16 - 17 | 2488 | 84 | |
| GenBank mRNA | AB100432.1 | - | 15 - 16 | 2282 | 84 | |
| AB100433.1 | - | 16 - 17 | 2294 | 84 | ||
| AF117610.1 | - | 16 - 17 | 2424 | 84 | ||
| AK034764.1 | - | 15 - 16 | 2748 | 84 | ||
| AK081841.1 | - | 16 - 17 | 2422 | 84 | ||
| AK088627.1 | - | 16 - 17 | 2445 | 84 | ||
| BC052414.1 | - | 16 - 17 | 2421 | 6401188 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Incenp
Genomic Map