Assay Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016692.3 | NP_057901.2 | 3 - 4 | 461 | 79 | |
| GenBank mRNA | AB100432.1 | - | 2 - 3 | 255 | 79 | |
| AB100433.1 | - | 2 - 3 | 255 | 79 | ||
| AF117610.1 | - | 3 - 4 | 397 | 79 | ||
| AK081841.1 | - | 3 - 4 | 395 | 79 | ||
| AK088627.1 | - | 3 - 4 | 418 | 79 | ||
| BC021761.2 | - | 3 - 4 | 358 | 79 | ||
| BC037011.1 | - | 3 - 4 | 414 | 79 | ||
| BC052414.1 | - | 3 - 4 | 394 | 6401188 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Incenp
Genomic Map