Assay Details
| Gene Symbol: Sema4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011976.1 | NP_036106.1 | 1 - 2 | 519 | 62 | |
| GenBank mRNA | AF134918.1 | - | 1 - 2 | 519 | 62 | |
| AK039355.1 | - | 2 - 3 | 603 | 62 | ||
| AK173219.1 | - | 1 - 2 | 3346 | 62 | ||
| BC049183.1 | - | 1 - 2 | 512 | 6395010 | 62 | |
| BC054532.1 | - | 2 - 3 | 557 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4g
Genomic Map