Assay Details
| Gene Symbol: Sema4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011976.1 | NP_036106.1 | 7 - 8 | 1199 | 108 | |
| GenBank mRNA | AF134918.1 | - | 7 - 8 | 1199 | 108 | |
| BC049183.1 | - | 7 - 8 | 1192 | 6395010 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema4g
Genomic Map