Assay Details
Gene Symbol: Zfp664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081750.1 | NP_001075219.1 | 2 - 3 | 331 | 100 | |
GenBank mRNA | AK035022.1 | - | 272 | 100 | ||
AK039671.1 | - | 2 - 3 | 332 | 100 | ||
AK086580.1 | - | 2 - 3 | 338 | 100 | ||
AK165974.1 | - | 2 - 3 | 294 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081750.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039038 | AK165974.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039038 | NM_001081750.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039038 | AK035022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039038 | AK086580.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083997 | BC096550.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083997 | BC027820.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083997 | NM_001081750.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652954 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591905 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725483 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510879 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113399 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384131 | AK008103.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731800 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13560 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121833 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195287 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131134 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126575 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50060 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126419 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100503 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177102 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083997 | BC096550.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083997 | BC027820.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083997 | NM_001081750.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039038 | AK165974.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039038 | NM_001081750.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039038 | AK035022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039038 | AK086580.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126419 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177102 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05731086 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05730999 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100503 | - | - |
Whole Mouse Genome G4122A | A_52_P529446 | AK165974.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P529446 | NM_001081750.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P529446 | AK086580.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P529446 | AK143192.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101913 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126419 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177102 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24063353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063279 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04787436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063300 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04779420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04787444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04135133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04168210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04787442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04703698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04923044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04356250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04168219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04809034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04534479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04534483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04484625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04472022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04484618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04168220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04696756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04484617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24063355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10525875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525878 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525873 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525879 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434063_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437047_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10525877 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10525872 | - | - |
Mouse Genome 430 2.0 Array | 1437047_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437047_at | - | - |
Mouse Genome 430 2.0 Array | 1434063_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1434063_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269704_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269704_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500013207.mm.1 | TC0500001453.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500013207.mm.1 | NM_001081750.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013207.mm.1 | NM_001081750.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013205.mm.1 | TC0500001453.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500013205.mm.1 | NM_001081750.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013205.mm.1 | NM_001081750.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013209.mm.1 | TC0500001453.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500013209.mm.1 | NM_001081750.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013209.mm.1 | NM_001081750.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013208.mm.1 | TC0500001453.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500013208.mm.1 | NM_001081750.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013208.mm.1 | NM_001081750.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013206.mm.1 | TC0500001453.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500013206.mm.1 | NM_001081750.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500013206.mm.1 | NM_001081750.1 | Refseq |
Mouse Expression Set 430 | 1434063_PM_at | - | - |
Mouse Expression Set 430 | 1434063_at | - | - |
Mouse Expression Set 430 | 1437047_at | - | - |
Mouse Expression Set 430 | 1437047_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437047_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434063_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434063_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437047_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 308395 | - | - |
ABI Mouse Genome Survey Microarray | 694788 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2525253 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2657340 | - | - |
