Assay Details
| Gene Symbol: Poll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020032.2 | NP_064416.1 | 5 - 6 | 970 | 87 | |
| GenBank mRNA | AF176099.1 | - | 4 - 5 | 911 | 87 | |
| AJ131889.1 | - | 5 - 6 | 970 | 87 | ||
| AK003392.1 | - | 1 - 2 | 106 | 87 | ||
| AK133087.1 | - | 5 - 6 | 939 | 87 | ||
| BC004767.1 | - | 4 - 5 | 920 | 3156259 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poll
Genomic Map