Assay Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009270.3 | NP_033296.1 | 1 - 2 | 1035 | 93 | |
| GenBank mRNA | AK138457.1 | - | 1 - 2 | 1046 | 93 | |
| AK148419.1 | - | 1 - 2 | 1051 | 93 | ||
| AK161837.1 | - | 1 - 2 | 1021 | 93 | ||
| AK163502.1 | - | 1 - 2 | 1015 | 93 | ||
| BC042781.1 | - | 1 - 2 | 292 | 4915911 | 93 | |
| BC056361.1 | - | 1 - 2 | 1035 | 6816800 | 93 | |
| D42048.1 | - | 1 - 2 | 901 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqle
Genomic Map