Assay Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009270.3 | NP_033296.1 | 6 - 7 | 1855 | 96 | |
| GenBank mRNA | AK138457.1 | - | 6 - 7 | 1866 | 96 | |
| AK148419.1 | - | 6 - 7 | 1840 | 96 | ||
| AK163502.1 | - | 6 - 7 | 1835 | 96 | ||
| AK177904.1 | - | 3 - 4 | 372 | 96 | ||
| AK189190.1 | - | 1 - 2 | 144 | 96 | ||
| AK210458.1 | - | 1 - 2 | 112 | 96 | ||
| BC042781.1 | - | 6 - 7 | 1112 | 4915911 | 96 | |
| BC056361.1 | - | 6 - 7 | 1855 | 6816800 | 96 | |
| D42048.1 | - | 6 - 7 | 1721 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqle
Genomic Map