Assay Details
| Gene Symbol: Aldh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035145.1 | - | 2 - 3 | 332 | 84 | |
| AK042326.1 | - | 2 - 3 | 336 | 84 | ||
| AK043506.1 | - | 2 - 3 | 326 | 84 | ||
| AK078626.1 | - | 2 - 3 | 332 | 84 | ||
| AK079602.1 | - | 2 - 3 | 307 | 84 | ||
| AK128913.1 | - | 2 - 3 | 269 | 84 | ||
| AK146286.1 | - | 2 - 3 | 325 | 84 | ||
| AK150233.1 | - | 2 - 3 | 324 | 84 | ||
| AK150250.1 | - | 2 - 3 | 337 | 84 | ||
| AK150920.1 | - | 2 - 3 | 324 | 84 | ||
| AK150986.1 | - | 2 - 3 | 306 | 84 | ||
| AK151763.1 | - | 2 - 3 | 324 | 84 | ||
| AK151826.1 | - | 2 - 3 | 306 | 84 | ||
| AK153128.1 | - | 2 - 3 | 306 | 84 | ||
| AK160060.1 | - | 2 - 3 | 326 | 84 | ||
| AK163452.1 | - | 332 | 84 | |||
| AK165032.1 | - | 2 - 3 | 570 | 84 | ||
| BC005476.1 | - | 2 - 3 | 314 | 3600875 | 84 | |
| S71509.1 | - | 2 - 3 | 291 | 84 | ||
| U07235.1 | - | 2 - 3 | 331 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh2
Genomic Map