Assay Details
| Gene Symbol: Brap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289543.1 | NP_001276472.1 | 9 - 10 | 1314 | 84 | |
| NM_001289544.1 | NP_001276473.1 | 10 - 11 | 1638 | 84 | ||
| NM_028227.3 | NP_082503.2 | 10 - 11 | 1510 | 84 | ||
| GenBank mRNA | AF321920.1 | - | 10 - 11 | 1402 | 84 | |
| AF321921.1 | - | 10 - 11 | 1639 | 84 | ||
| AK013885.1 | - | 8 - 9 | 1003 | 84 | ||
| AK034212.1 | - | 5 - 6 | 443 | 84 | ||
| AK143718.1 | - | 9 - 10 | 1304 | 84 | ||
| AK177250.1 | - | 1 - 2 | 52 | 84 | ||
| BC038490.1 | - | 7 - 8 | 879 | 3599718 | 84 | |
| BC062932.1 | - | 10 - 11 | 1486 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brap
Genomic Map