Assay Details
| Gene Symbol: Pyroxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029011.2 | NP_083287.2 | 11 - 12 | 1446 | 62 | |
| GenBank mRNA | AK154222.1 | - | 11 - 12 | 1208 | 62 | |
| AK160239.1 | - | 11 - 12 | 1163 | 62 | ||
| BC140971.1 | - | 11 - 12 | 1275 | 9055583 | 62 | |
| BC144951.1 | - | 11 - 12 | 1275 | 9053499 | 62 | |
| CX218741.1 | - | 4 - 5 | 392 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pyroxd2
Genomic Map