Assay Details
| Gene Symbol: Pyroxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029011.2 | NP_083287.2 | 5 - 6 | 784 | 78 | |
| GenBank mRNA | AK160239.1 | - | 5 - 6 | 500 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pyroxd2
Genomic Map