Assay Details
Gene Symbol: 2410131K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081236.1 | NP_001074705.1 | 1 - 2 | 237 | 62 | |
GenBank mRNA | AK010791.1 | - | 1 - 2 | 214 | 62 | |
AK090380.1 | - | 1 - 2 | 196 | 62 | ||
BC104372.1 | - | 1 - 2 | 171 | 40049898 | 62 | |
BC104373.1 | - | 1 - 2 | 171 | 40049899 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
