Assay Details
Gene Symbol: 2410131K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC104372.1 | - | 2 - 3 | 323 | 40049898 | 72 |
BC104373.1 | - | 2 - 3 | 323 | 40049899 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
