Assay Details
Gene Symbol: 2410131K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081236.1 | NP_001074705.1 | 3 - 4 | 544 | 85 | |
GenBank mRNA | AK010791.1 | - | 3 - 4 | 521 | 85 | |
AK090380.1 | - | 3 - 4 | 503 | 85 | ||
BC104372.1 | - | 3 - 4 | 478 | 40049898 | 85 | |
BC104373.1 | - | 3 - 4 | 478 | 40049899 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
