Assay Details
| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083925.1 | NP_001077394.1 | 2 - 3 | 521 | 74 | |
| NR_003507.1 | 2 - 3 | 524 | 74 | |||
| GenBank mRNA | AB067529.1 | - | 2 - 3 | 524 | 74 | |
| AF328926.1 | - | 2 - 3 | 521 | 74 | ||
| AF418004.1 | - | 2 - 3 | 521 | 74 | ||
| AF418005.1 | - | 2 - 3 | 521 | 74 | ||
| AF466822.1 | - | 2 - 3 | 478 | 74 | ||
| AK080152.1 | - | 2 - 3 | 514 | 74 | ||
| X55982.1 | - | 1 - 2 | 287 | 74 | ||
More Information
Set Membership: |
|

Oas1b
Genomic Map