Assay Details
| Gene Symbol: Slc22a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC026439.1 | - | 1 - 2 | 73 | 4239636 | 96 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026439.1 transcripts.
Set Membership: |
|

Slc22a22
Genomic Map