Assay Details
| Gene Symbol: Slc22a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172378.2 | NP_759010.1 | 3 - 4 | 841 | 99 | |
| GenBank mRNA | AB520670.1 | - | 2 - 3 | 447 | 99 | |
| AK030222.1 | - | 3 - 4 | 841 | 99 | ||
| BC026439.1 | - | 2 - 3 | 484 | 4239636 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a22
Genomic Map