Assay Details
| Gene Symbol: Slc22a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172378.2 | NP_759010.1 | 5 - 6 | 1103 | 126 | |
| GenBank mRNA | AB520670.1 | - | 4 - 5 | 709 | 126 | |
| AK030222.1 | - | 5 - 6 | 1103 | 126 | ||
| BC026439.1 | - | 4 - 5 | 746 | 4239636 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a22
Genomic Map