Assay Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058275.1 | - | 1 - 2 | 183 | 6515674 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfrl| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058275.1 | - | 1 - 2 | 183 | 6515674 | 77 |
Set Membership: |
|