Assay Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026840.3 | NP_081116.3 | 3 - 4 | 649 | 71 | |
| GenBank mRNA | AK004179.1 | - | 3 - 4 | 541 | 71 | |
| AK132625.1 | - | 3 - 4 | 650 | 71 | ||
| BC058275.1 | - | 3 - 4 | 641 | 6515674 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfrl
Genomic Map