Assay Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026840.3 | NP_081116.3 | 4 - 5 | 942 | 70 | |
| GenBank mRNA | AK004179.1 | - | 4 - 5 | 834 | 70 | |
| AK132625.1 | - | 4 - 5 | 943 | 70 | ||
| BC039803.1 | - | 2 - 3 | 326 | 70 | ||
| BC058275.1 | - | 4 - 5 | 934 | 6515674 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfrl
Genomic Map