Assay Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199011.1 | NP_950176.1 | 14 - 15 | 1672 | 69 | |
| GenBank mRNA | AK043933.1 | - | 1386 | 69 | ||
| AK048764.1 | - | 14 - 15 | 1714 | 69 | ||
| AK132151.1 | - | 13 - 14 | 1579 | 69 | ||
| AK170840.1 | - | 13 - 14 | 1768 | 69 | ||
| BC062929.1 | - | 14 - 15 | 1672 | 6855058 | 69 | |
| BC066012.1 | - | 14 - 15 | 1649 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkq
Genomic Map