Assay Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199011.1 | NP_950176.1 | 15 - 16 | 1783 | 74 | |
| GenBank mRNA | AK043933.1 | - | 1497 | 74 | ||
| AK132151.1 | - | 14 - 15 | 1690 | 74 | ||
| AK134745.1 | - | 2 - 3 | 846 | 74 | ||
| BC062929.1 | - | 15 - 16 | 1783 | 6855058 | 74 | |
| BC066012.1 | - | 15 - 16 | 1760 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkq
Genomic Map