Assay Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199011.1 | NP_950176.1 | 1 - 2 | 324 | 79 | |
| GenBank mRNA | AK048764.1 | - | 1 - 2 | 366 | 79 | |
| AK132151.1 | - | 1 - 2 | 367 | 79 | ||
| AK170840.1 | - | 1 - 2 | 345 | 79 | ||
| BC062929.1 | - | 1 - 2 | 324 | 6855058 | 79 | |
| BC066012.1 | - | 1 - 2 | 347 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkq
Genomic Map