Assay Details
| Gene Symbol: Dgkq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199011.1 | NP_950176.1 | 4 - 5 | 586 | 59 | |
| GenBank mRNA | AK043933.1 | - | 436 | 59 | ||
| AK048764.1 | - | 4 - 5 | 628 | 59 | ||
| AK132151.1 | - | 4 - 5 | 583 | 59 | ||
| AK170840.1 | - | 4 - 5 | 607 | 59 | ||
| BC062929.1 | - | 4 - 5 | 586 | 6855058 | 59 | |
| BC066012.1 | - | 4 - 5 | 563 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkq
Genomic Map