Assay Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008325.4 | NP_032351.2 | 13 - 14 | 2148 | 121 | |
| NM_172997.1 | NP_766585.1 | 12 - 13 | 2007 | 121 | ||
| GenBank mRNA | AK031414.1 | - | 12 - 13 | 1802 | 121 | |
| AK045330.1 | - | 11 - 12 | 1635 | 121 | ||
| AK155627.1 | - | 13 - 14 | 2149 | 121 | ||
| AK169998.1 | - | 12 - 13 | 1806 | 121 | ||
| AK171124.1 | - | 8 - 9 | 1158 | 121 | ||
| BC057935.1 | - | 12 - 13 | 1769 | 5327150 | 121 | |
| L34111.1 | - | 12 - 13 | 1722 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idua
Genomic Map