Assay Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008325.4 | NP_032351.2 | 4 - 5 | 806 | 68 | |
| NM_172997.1 | NP_766585.1 | 3 - 4 | 665 | 68 | ||
| GenBank mRNA | AK031414.1 | - | 3 - 4 | 460 | 68 | |
| AK045330.1 | - | 2 - 3 | 293 | 68 | ||
| AK155627.1 | - | 4 - 5 | 807 | 68 | ||
| AK169998.1 | - | 3 - 4 | 464 | 68 | ||
| BC057935.1 | - | 3 - 4 | 427 | 5327150 | 68 | |
| L34111.1 | - | 3 - 4 | 380 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idua
Genomic Map