Assay Details
| Gene Symbol: Mfsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172883.2 | NP_766471.1 | 2 - 3 | 333 | 83 | |
| GenBank mRNA | AK029021.1 | - | 2 - 3 | 333 | 83 | |
| AK087226.1 | - | 2 - 3 | 543 | 83 | ||
| AK171069.1 | - | 1 - 2 | 2965 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd7a
Genomic Map