Assay Details
| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164259.1 | NP_001157731.1 | 4 - 5 | 899 | 93 | |
| NM_054071.2 | NP_473412.1 | 5 - 6 | 1172 | 93 | ||
| GenBank mRNA | AF321300.1 | - | 5 - 6 | 899 | 93 | |
| AF321301.1 | - | 4 - 5 | 860 | 93 | ||
| AJ293947.1 | - | 4 - 5 | 805 | 93 | ||
| BC058745.1 | - | 4 - 5 | 774 | 6439421 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgfrl1
Genomic Map