Assay Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163531.1 | NP_001157003.1 | 5 - 6 | 531 | 89 | |
| NM_001163532.1 | NP_001157004.1 | 5 - 6 | 534 | 89 | ||
| NM_028223.3 | NP_082499.3 | 5 - 6 | 641 | 89 | ||
| GenBank mRNA | AI526594.1 | - | 5 - 6 | 505 | 89 | |
| AK013879.1 | - | 5 - 6 | 464 | 89 | ||
| AK034579.1 | - | 5 - 6 | 510 | 89 | ||
| AK038865.1 | - | 5 - 6 | 501 | 89 | ||
| AK049952.1 | - | 5 - 6 | 575 | 89 | ||
| AK082765.1 | - | 5 - 6 | 497 | 89 | ||
| AK170273.1 | - | 5 - 6 | 484 | 89 | ||
| BB653267.1 | - | 5 - 6 | 571 | 89 | ||
| BC005542.1 | - | 5 - 6 | 449 | 3590497 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem175
Genomic Map