Assay Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172716.4 | NP_766304.2 | 5 - 6 | 521 | 66 | |
| GenBank mRNA | AK033609.1 | - | 5 - 6 | 596 | 66 | |
| AK085574.1 | - | 5 - 6 | 577 | 66 | ||
| AK089091.1 | - | 5 - 6 | 522 | 66 | ||
| BC049266.1 | - | 388 | 5371040 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf3
Genomic Map