Assay Details
| Gene Symbol: 2810404M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045497.1 | 5 - 6 | 639 | 115 | ||
| GenBank mRNA | AK012985.1 | - | 5 - 6 | 639 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FSHD region gene 1 | Frg1 | -627.5kb | Anti-sense | Chr. 8 - 41397454 - 41417118 on NCBI Build GRCm38 | |
| Downstream |
zinc finger protein 353, pseudogene |
Zfp353-ps | Zfp353 | 35.1kb | Anti-sense | Chr. 8 - 42081329 - 42083312 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2279777 | AK012985.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07126289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07394703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07127446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07140880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07318117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07318109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06981243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07174327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25813291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25812862 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1453940_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453940_at | - | - |
| Mouse Genome 430 2.0 Array | 1453940_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800003454.mm.1 | NR_045497.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800003454.mm.1 | NR_045497.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800003454.mm.1 | TC0800000496.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453940_PM_at | - | - |
| Mouse Expression Set 430 | 1453940_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453940_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453940_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 712150 | - | - |

2810404M03Rik
Genomic Map