Assay Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 11 - 12 | 1338 | 129 | |
| GenBank mRNA | AK087867.1 | - | 5 - 6 | 611 | 129 | |
| AK088543.1 | - | 13 - 14 | 2580 | 129 | ||
| AK139654.1 | - | 11 - 12 | 1339 | 129 | ||
| BC037640.1 | - | 11 - 12 | 1316 | 5096937 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l1
Genomic Map