Assay Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 2 - 3 | 193 | 122 | |
| GenBank mRNA | AK088543.1 | - | 4 - 5 | 1435 | 122 | |
| AK139654.1 | - | 2 - 3 | 194 | 122 | ||
| BC027553.1 | - | 2 - 3 | 178 | 122 | ||
| BC037640.1 | - | 2 - 3 | 171 | 5096937 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088543.1 transcripts.
Set Membership: |
|

Cwf19l1
Genomic Map