Assay Details
| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 6 - 7 | 715 | 86 | |
| GenBank mRNA | AK012163.1 | - | 6 - 7 | 699 | 86 | |
| AK088543.1 | - | 8 - 9 | 1957 | 86 | ||
| AK139654.1 | - | 6 - 7 | 716 | 86 | ||
| BC027553.1 | - | 6 - 7 | 700 | 86 | ||
| BC037640.1 | - | 6 - 7 | 693 | 5096937 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l1
Genomic Map