Assay Details
| Gene Symbol: Gsdmcl-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029414.1 | 71 | 60 | |||
| GenBank mRNA | AK016931.1 | - | 72 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gsdmcl-ps
Genomic Map