Assay Details
| Gene Symbol: Cutc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113562.1 | NP_001107034.1 | 7 - 8 | 736 | 74 | |
| NM_025530.3 | NP_079806.2 | 7 - 8 | 706 | 74 | ||
| GenBank mRNA | AK007591.1 | - | 7 - 8 | 661 | 74 | |
| AK009702.1 | - | 7 - 8 | 731 | 74 | ||
| BC026775.1 | - | 7 - 8 | 646 | 3582902 | 74 | |
| BP754125.1 | - | 2 - 3 | 74 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cutc
Genomic Map